Anti-DIABLO Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: DIABLO
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-451
- 77836-450
- Antigen Symbol:
- DIABLO
- DIABLO
- Antigen Name:
- Diablo homolog, mitochondrial protein (168-236AA)
- Diablo homolog, mitochondrial protein (168-236AA)
- Antigen Synonyms:
- 0610041G12Rik|DBLOH_HUMAN|DBOH|DFNA64|Diablo homolog (Drosophila)|Diablo homolog|Diablo homolog mitochondrial|Diablo IAP binding mitochondrial protein|Diablo like protein|DIABLO S|Direct IAP binding protein with low pI|Direct IAP-binding protein with low pI|FLJ10537|FLJ25049|mitochondrial|Mitochondrial Smac protein|Second mitochondria derived activator of caspase|Second mitochondria-derived activator of caspase|second mitochondrial activator of caspases|SMAC 3|Smac|Smac protein|SMAC3
- 0610041G12Rik|DBLOH_HUMAN|DBOH|DFNA64|Diablo homolog (Drosophila)|Diablo homolog|Diablo homolog mitochondrial|Diablo IAP binding mitochondrial protein|Diablo like protein|DIABLO S|Direct IAP binding protein with low pI|Direct IAP-binding protein with low pI|FLJ10537|FLJ25049|mitochondrial|Mitochondrial Smac protein|Second mitochondria derived activator of caspase|Second mitochondria-derived activator of caspase|second mitochondrial activator of caspases|SMAC 3|Smac|Smac protein|SMAC3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NR28
- Q9NR28
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together