Anti-NAA15 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: NAA15
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-926
- 77835-927
- Antigen Symbol:
- NAA15
- NAA15
- Antigen Name:
- N-alpha-acetyltransferase 15, NatA auxiliary subunit protein (742-865AA)
- N-alpha-acetyltransferase 15, NatA auxiliary subunit protein (742-865AA)
- Antigen Synonyms:
- 5730450D16Rik|6330400I15|ASTBDN|GA19|Gastric cancer antigen Ga19|MGC29428|mNAT1|N alpha acetyltransferase 15, NatA auxiliary subunit|N terminal acetyltransferase|N(alpha) acetyltransferase 15, NatA auxiliary subunit|N-alpha-acetyltransferase 15|N-terminal acetyltransferase|Naa15|NAA15_HUMAN|NatA auxiliary subunit|NATH|NMDA receptor regulated 1|NMDA receptor regulated protein 1|NMDA receptor-regulated protein 1|Protein tubedown 1|Protein tubedown-1|Tbdn 1|Tbdn100|Transcriptional coactivator tubedown 100|tubedown-1
- 5730450D16Rik|6330400I15|ASTBDN|GA19|Gastric cancer antigen Ga19|MGC29428|mNAT1|N alpha acetyltransferase 15, NatA auxiliary subunit|N terminal acetyltransferase|N(alpha) acetyltransferase 15, NatA auxiliary subunit|N-alpha-acetyltransferase 15|N-terminal acetyltransferase|Naa15|NAA15_HUMAN|NatA auxiliary subunit|NATH|NMDA receptor regulated 1|NMDA receptor regulated protein 1|NMDA receptor-regulated protein 1|Protein tubedown 1|Protein tubedown-1|Tbdn 1|Tbdn100|Transcriptional coactivator tubedown 100|tubedown-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXJ9
- Q9BXJ9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together