Anti-BATF Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: BATF
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-757
- 77823-756
- Antigen Symbol:
- BATF
- BATF
- Antigen Name:
- Basic leucine zipper transcriptional factor ATF-like protein (1-125AA)
- Basic leucine zipper transcriptional factor ATF-like protein (1-125AA)
- Antigen Synonyms:
- Activating transcription factor B|B ATF|B-ATF|B-cell-activating transcription factor|Basic leucine zipper transcription factor like|Basic leucine zipper transcriptional factor ATF like|Basic leucine zipper transcriptional factor ATF-like|Batf|BATF_HUMAN|BATF1|SF HT activated gene 2 protein|SF-HT-activated gene 2 protein|SFA 2|SFA-2|SFA2
- Activating transcription factor B|B ATF|B-ATF|B-cell-activating transcription factor|Basic leucine zipper transcription factor like|Basic leucine zipper transcriptional factor ATF like|Basic leucine zipper transcriptional factor ATF-like|Batf|BATF_HUMAN|BATF1|SF HT activated gene 2 protein|SF-HT-activated gene 2 protein|SFA 2|SFA-2|SFA2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16520
- Q16520
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together