Anti-DDX50 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DDX50
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-172
- 77835-171
- Antigen Symbol:
- DDX50
- DDX50
- Antigen Name:
- ATP-dependent RNA helicase DDX50 protein (1-210AA)
- ATP-dependent RNA helicase DDX50 protein (1-210AA)
- Antigen Synonyms:
- 4933429B04Rik|ATP-dependent RNA helicase |Ddx50|_HUMAN|DEAD (Asp-Glu-Ala-Asp) box polypeptide 50|DEAD box protein 50|Gu beta|Gu-beta|GU2|GUB|MGC109605|MGC3199|Nucleolar protein Gu2|RH II|RH II/GuB|RNA helicase II / Gu beta
- 4933429B04Rik|ATP-dependent RNA helicase |Ddx50|_HUMAN|DEAD (Asp-Glu-Ala-Asp) box polypeptide 50|DEAD box protein 50|Gu beta|Gu-beta|GU2|GUB|MGC109605|MGC3199|Nucleolar protein Gu2|RH II|RH II/GuB|RNA helicase II / Gu beta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQ39
- Q9BQ39
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together