Anti-FBXW12 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FBXW12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-416
- 77830-415
- Antigen Symbol:
- FBXW12
- FBXW12
- Antigen Name:
- F-box/WD repeat-containing protein 12 protein (1-250AA)
- F-box/WD repeat-containing protein 12 protein (1-250AA)
- Antigen Synonyms:
- FBXW12|FBW12|FBXO12|FBXO35F-box/WD repeat-containing protein 12|F-box and WD-40 domain-containing protein 12|F-box only protein 35
- FBXW12|FBW12|FBXO12|FBXO35F-box/WD repeat-containing protein 12|F-box and WD-40 domain-containing protein 12|F-box only protein 35
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6X9E4
- Q6X9E4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together