Anti-MBTPS2 Rabbit Polyclonal Antibody
77798-012
New Product
- Antigen Name:Synthesized peptide derived from the Internal region of S2P.
- Antigen Symbol:MBTPS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:O43462
- Antigen Synonyms:BRESEK|Endopeptidase S2P|IFAP|KFSDX|MBTP2_HUMAN|MBTPS2|Membrane bound transcription factor site 2 protease|membrane-bound transcription factor peptidase, site 2|membrane-bound transcription factor protease, site 2|Membrane-bound transcription factor site-2 protease|S2P|S2P endopeptidase|Site 2 protease|SREBPs intramembrane protease|Sterol regulatory element-binding proteins intramembrane protease
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IF:1:200 to 1:1000, ELISA:1:20000
Type:
Antigen: MBTPS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat