Anti-PIGU Rabbit Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIGU
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-024
- 77842-025
- Antigen Symbol:
- PIGU
- PIGU
- Antigen Name:
- Phosphatidylinositol glycan anchor biosynthesis class U protein (28-47AA)
- Phosphatidylinositol glycan anchor biosynthesis class U protein (28-47AA)
- Antigen Synonyms:
- 5430426F17Rik|BC028278|Cdc91l1|Cell division cycle 91 like 1 protein|Cell division cycle protein 91-like 1|GAB1|GPI transamidase component PIG-U|GPI transamidase subunit|Phosphatidylinositol glycan anchor biosynthesis class U|Phosphatidylinositol glycan anchor biosynthesis class U protein|Pigu|PIGU_HUMAN|Protein CDC91 like 1|Protein CDC91-like 1
- 5430426F17Rik|BC028278|Cdc91l1|Cell division cycle 91 like 1 protein|Cell division cycle protein 91-like 1|GAB1|GPI transamidase component PIG-U|GPI transamidase subunit|Phosphatidylinositol glycan anchor biosynthesis class U|Phosphatidylinositol glycan anchor biosynthesis class U protein|Pigu|PIGU_HUMAN|Protein CDC91 like 1|Protein CDC91-like 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H490
- Q9H490
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together