Anti-RFWD3 Rabbit Polyclonal Antibody
Catalog # 77807-545
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of RFWD3.
- Antigen Symbol:RFWD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6PCD5
- Antigen Synonyms:BC027246|E3 ubiquitin protein ligase RFWD3|E3 ubiquitin-protein ligase RFWD3|FLJ10520|MGC27888|RFWD3|RFWD3_HUMAN|Ring finger and WD repeat domain 3|RING finger and WD repeat domain containing protein 3|RING finger and WD repeat domain-containing protein 3|RING finger protein 201|RNF201
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: RFWD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human