Anti-SALL2 Rabbit Polyclonal Antibody
Catalog # 77798-637
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:hSal2
- Antigen Symbol:SALL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y467
- Antigen Synonyms:SALL2|KIAA0360|SAL2|ZNF795|Sal-like protein 2|Zinc finger protein 795|Zinc finger protein SALL2|Zinc finger protein Spalt-2|Sal-2|hSal2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SALL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat