Anti-SLC25A4 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SLC25A4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-914
- 77799-913
- Antigen Symbol:
- SLC25A4
- SLC25A4
- Antigen Name:
- ADP/ATP translocase 1 protein (40-74AA)
- ADP/ATP translocase 1 protein (40-74AA)
- Antigen Synonyms:
- AAC1|Adenine Nucleotide Translocase 1|Adenine nucleotide translocator 1 (skeletal muscle)|Adenine nucleotide translocator 1|ADP|ADP ATP carrier protein 1|ADP ATP carrier protein heart/skeletal muscle isoform T1|ADP/ATP translocase 1|ADT1_HUMAN|ANT 1|ANT|ANT1|ATP carrier protein 1|ATP carrier protein|heart/skeletal muscle isoform T1|PEO2|PEO3|SLC25A4|Solute carrier family 25 member 4|T1
- AAC1|Adenine Nucleotide Translocase 1|Adenine nucleotide translocator 1 (skeletal muscle)|Adenine nucleotide translocator 1|ADP|ADP ATP carrier protein 1|ADP ATP carrier protein heart/skeletal muscle isoform T1|ADP/ATP translocase 1|ADT1_HUMAN|ANT 1|ANT|ANT1|ATP carrier protein 1|ATP carrier protein|heart/skeletal muscle isoform T1|PEO2|PEO3|SLC25A4|Solute carrier family 25 member 4|T1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12235
- P12235
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together