Anti-USP32 Rabbit Polyclonal Antibody
Catalog # 77804-139
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Deubiquitinating enzyme 32
- Antigen Symbol:USP32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NFA0
- Antigen Synonyms:Deubiquitinating enzyme 32|NY REN 60|NYREN60|Renal carcinoma antigen NY REN 60|Renal carcinoma antigen NY-REN-60|Ubiquitin carboxyl-terminal hydrolase 32|Ubiquitin specific peptidase 32|Ubiquitin specific processing protease 32|Ubiquitin specific protease 32|Ubiquitin thioesterase 32|Ubiquitin thiolesterase 32|Ubiquitin-specific-processing protease 32|UBP32_HUMAN|USP10|USP32
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: USP32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse