Anti-NEDD9 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NEDD9
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-851
- 77824-852
- Antigen Symbol:
- NEDD9
- NEDD9
- Antigen Name:
- Enhancer of filamentation 1 protein (68-151AA)
- Enhancer of filamentation 1 protein (68-151AA)
- Antigen Synonyms:
- Cas like docking|Cas scaffolding protein family member 2|CAS-L|CAS2|CasL|CASL_HUMAN|CASS2|Crk associated substrate related|Crk associated substrate related protein|CRK-associated substrate-related protein|dJ49G10.2 (Enhancer of Filamentation 1 (HEF1))|dJ49G10.2|dJ761I2.1 (enhancer of filamentation (HEF1))|dJ761I2.1|Enhancer of filamentation 1|Enhancer of filamentation 1 p55|HEF 1|HEF1|NEDD-9|Nedd9|Neural precursor cell expressed developmentally down-regulated protein 9|P105|Renal carcinoma antigen NY-REN-12
- Cas like docking|Cas scaffolding protein family member 2|CAS-L|CAS2|CasL|CASL_HUMAN|CASS2|Crk associated substrate related|Crk associated substrate related protein|CRK-associated substrate-related protein|dJ49G10.2 (Enhancer of Filamentation 1 (HEF1))|dJ49G10.2|dJ761I2.1 (enhancer of filamentation (HEF1))|dJ761I2.1|Enhancer of filamentation 1|Enhancer of filamentation 1 p55|HEF 1|HEF1|NEDD-9|Nedd9|Neural precursor cell expressed developmentally down-regulated protein 9|P105|Renal carcinoma antigen NY-REN-12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14511
- Q14511
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together