Anti-PABPC4 Rabbit Polyclonal Antibody
Catalog # 77793-099
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Activated platelet protein 1
- Antigen Symbol:PABPC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13310
- Antigen Synonyms:Activated-platelet protein 1|APP 1 |APP-1|APP1 |FLJ43938|Inducible poly(A) binding protein|Inducible poly(A)-binding protein|iPABP|IPABP4|PABP 4|PABP-4|PABP4|PABP4_HUMAN|PABPC 4|PABPC4|Poly A binding protein cytoplasmic 4|Poly(A) binding protein 4|Poly(A) binding protein cytoplasmic 4 (inducible form) |Poly(A) binding protein cytoplasmic 4|Poly(A)-binding protein 4|Polyadenylate binding protein 4|Polyadenylate-binding protein 4|Polyadenylate-binding protein|cytoplasmic|4|inducible
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PABPC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat