Anti-VTI1B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VTI1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-690
- 77842-689
- Antigen Symbol:
- VTI1B
- VTI1B
- Antigen Name:
- Vesicle transport through interaction with t-SNAREs homolog 1B protein (2-208AA)
- Vesicle transport through interaction with t-SNAREs homolog 1B protein (2-208AA)
- Antigen Synonyms:
- HGNC:17793|V SNARE|v-SNARE|Vesicle associated soluble NSF attachment protein receptor|Vesicle transport through interaction with t SNAREs 1B|Vesicle transport through interaction with t SNAREs homolog 1B (yeast)|Vesicle transport through interaction with t-SNAREs homolog 1B|Vesicle transport v-SNARE protein Vti1-like 1|VTI1|VTI1 like|Vti1 rp1|VTI1-LIKE|Vti1-rp1|VTI1B|VTI1B_HUMAN|VTI1L|VTI1L1|VTI2
- HGNC:17793|V SNARE|v-SNARE|Vesicle associated soluble NSF attachment protein receptor|Vesicle transport through interaction with t SNAREs 1B|Vesicle transport through interaction with t SNAREs homolog 1B (yeast)|Vesicle transport through interaction with t-SNAREs homolog 1B|Vesicle transport v-SNARE protein Vti1-like 1|VTI1|VTI1 like|Vti1 rp1|VTI1-LIKE|Vti1-rp1|VTI1B|VTI1B_HUMAN|VTI1L|VTI1L1|VTI2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UEU0
- Q9UEU0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together