Anti-LAMTOR1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LAMTOR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-820
- 77830-821
- Antigen Symbol:
- LAMTOR1
- LAMTOR1
- Antigen Name:
- Ragulator complex protein LAMTOR1 protein (2-161AA)
- Ragulator complex protein LAMTOR1 protein (2-161AA)
- Antigen Synonyms:
- C11orf59| LAMTOR1| Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1| Lipid raft adaptor protein p18| LTOR1_HUMAN| p18| p27Kip1-releasing factor from RhoA| p27RF-Rho| PDRO| PP7157| Protein associated with DRMs and endosomes| Ragulator complex protein LAMTOR1| ragulator complex protein PDRO| Ragulator1| RhoA activator C11orf59
- C11orf59| LAMTOR1| Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1| Lipid raft adaptor protein p18| LTOR1_HUMAN| p18| p27Kip1-releasing factor from RhoA| p27RF-Rho| PDRO| PP7157| Protein associated with DRMs and endosomes| Ragulator complex protein LAMTOR1| ragulator complex protein PDRO| Ragulator1| RhoA activator C11orf59
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6IAA8
- Q6IAA8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together