Anti-PRB1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-233
- 77795-232
- Antigen Symbol:
- PRB1
- PRB1
- Antigen Name:
- Basic salivary proline-rich protein 1 protein (17-92AA)
- Basic salivary proline-rich protein 1 protein (17-92AA)
- Antigen Synonyms:
- PRB1|Basic salivary proline-rich protein 1|Salivary proline-rich protein) [Cleaved into: Proline-rich peptide II-2|Basic peptide IB-6|Peptide P-H]
- PRB1|Basic salivary proline-rich protein 1|Salivary proline-rich protein) [Cleaved into: Proline-rich peptide II-2|Basic peptide IB-6|Peptide P-H]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04280
- P04280
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together