Anti-CYP27A1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: CYP27A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77806-715
- 77806-714
- Antigen Symbol:
- CYP27A1
- CYP27A1
- Antigen Name:
- Fusion protein of CYP27A1
- Fusion protein of CYP27A1
- Antigen Synonyms:
- 12-alpha-triol 27-hydroxylase|5-beta-cholestane-3-alpha|7-alpha|12-alpha-triol 26-hydroxylase|5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase|Cholestanetriol 26 monooxygenase|CP27|CP27A_HUMAN|CTX|CYP|CYP27|CYP27A1|Cytochrome P 450C27/25|Cytochrome P-450C27/25|Cytochrome P450 27|Cytochrome P450 27 mitochondrial|Cytochrome P450 family 27 subfamily A member 1|Cytochrome P450 family 27 subfamily A polypeptide 1|Cytochrome P450 subfamily XXVIIA (steroid 27-hydroxylase cerebrotendinous xanthomatosis) polypeptide 1|mitochondrial|Sterol 26 hydroxylase|Sterol 26 hydroxylase mitochondrial|Sterol 26-hydroxylase|Sterol 27 hydroxylase|Sterol 27-hydroxylase|Vitamin D(3) 25 hydroxylase|Vitamin D(3) 25-hydroxylase
- 12-alpha-triol 27-hydroxylase|5-beta-cholestane-3-alpha|7-alpha|12-alpha-triol 26-hydroxylase|5-beta-cholestane-3-alpha,7-alpha,12-alpha-triol 27-hydroxylase|Cholestanetriol 26 monooxygenase|CP27|CP27A_HUMAN|CTX|CYP|CYP27|CYP27A1|Cytochrome P 450C27/25|Cytochrome P-450C27/25|Cytochrome P450 27|Cytochrome P450 27 mitochondrial|Cytochrome P450 family 27 subfamily A member 1|Cytochrome P450 family 27 subfamily A polypeptide 1|Cytochrome P450 subfamily XXVIIA (steroid 27-hydroxylase cerebrotendinous xanthomatosis) polypeptide 1|mitochondrial|Sterol 26 hydroxylase|Sterol 26 hydroxylase mitochondrial|Sterol 26-hydroxylase|Sterol 27 hydroxylase|Sterol 27-hydroxylase|Vitamin D(3) 25 hydroxylase|Vitamin D(3) 25-hydroxylase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02318
- Q02318
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together