Anti-USP37 Rabbit Polyclonal Antibody
100 μl, Vial
Catalog # 77804-143
Supplier: AFG Bioscience
Supplier #: A38165-100UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: USP37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 μl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77804-143
- Reactivity:Human|Mouse|Rat
- Western Blot:Yes
- Shipping Temperature:Shipped on Blue ice
- Antigen Symbol:USP37
- Gene ID:Q86T82
- Conjugation:Unconjugated
- Antigen Synonyms:Deubiquitinating enzyme 37|EC 3.1.2.15|KIAA1594|MGC117261|OTTHUMP00000164146|tmp_locus_50|Ubiquitin carboxyl-terminal hydrolase 37|Ubiquitin specific peptidase 37|Ubiquitin specific processing protease 37|Ubiquitin specific protease 37|Ubiquitin thioesterase 37|Ubiquitin thiolesterase 37|Ubiquitin-specific-processing protease 37|UBP37_HUMAN
- Antigen Name:Deubiquitinating enzyme 3
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



