Anti-SND1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SND1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-882
- 77829-883
- Antigen Symbol:
- SND1
- SND1
- Antigen Name:
- Staphylococcal nuclease domain-containing protein 1 protein (560-732AA)
- Staphylococcal nuclease domain-containing protein 1 protein (560-732AA)
- Antigen Synonyms:
- 100 kDa coactivator|EBNA 2 co activator (100kD)|EBNA2 coactivator p100|p100|p100 co-activator|p100 coactivator|p100 EBNA2 co activator|SND 1|Snd1|_HUMAN|Staphylococcal nuclease and tudor domain containing 1|Staphylococcal nuclease domain containing 1|Staphylococcal nuclease domain containing protein 1|Staphylococcal nuclease domain-containing protein 1|TDRD11|TRD 11|TRD11|Tudor domain containing protein 11|Tudor domain-containing protein 11
- 100 kDa coactivator|EBNA 2 co activator (100kD)|EBNA2 coactivator p100|p100|p100 co-activator|p100 coactivator|p100 EBNA2 co activator|SND 1|Snd1|_HUMAN|Staphylococcal nuclease and tudor domain containing 1|Staphylococcal nuclease domain containing 1|Staphylococcal nuclease domain containing protein 1|Staphylococcal nuclease domain-containing protein 1|TDRD11|TRD 11|TRD11|Tudor domain containing protein 11|Tudor domain-containing protein 11
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7KZF4
- Q7KZF4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together