Anti-CAB39 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: CAB39
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-716
- 77825-717
- Antigen Symbol:
- CAB39
- CAB39
- Antigen Name:
- Calcium-binding protein 39 protein (28-290AA)
- Calcium-binding protein 39 protein (28-290AA)
- Antigen Synonyms:
- Cab39|CAB39_HUMAN|Calcium binding protein 39|Calcium-binding protein 39|CGI 66|CGI-66|CGI66|MO25 alpha|MO25|Mouse protein 25 alpha|Protein Mo25
- Cab39|CAB39_HUMAN|Calcium binding protein 39|Calcium-binding protein 39|CGI 66|CGI-66|CGI66|MO25 alpha|MO25|Mouse protein 25 alpha|Protein Mo25
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y376
- Q9Y376
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together