Anti-LYSMD3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LYSMD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-810
- 77831-809
- Antigen Symbol:
- LYSMD3
- LYSMD3
- Antigen Name:
- LysM and putative peptidoglycan-binding domain-containing protein 3 protein (1-127AA)
- LysM and putative peptidoglycan-binding domain-containing protein 3 protein (1-127AA)
- Antigen Synonyms:
- 1110030H10Rik|BC003322|lysM and putative peptidoglycan binding domain containing protein 3|LysM and putative peptidoglycan-binding domain-containing protein 3|LYSM3_HUMAN|lysmd3|RGD1308805
- 1110030H10Rik|BC003322|lysM and putative peptidoglycan binding domain containing protein 3|LysM and putative peptidoglycan-binding domain-containing protein 3|LYSM3_HUMAN|lysmd3|RGD1308805
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z3D4
- Q7Z3D4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together