Anti-ACAD11 Rabbit Polyclonal Antibody
Catalog # 77810-249
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of ACAD-11.
- Antigen Symbol:ACAD11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q709F0
- Antigen Synonyms:5730439E10Rik|62113 protein|ACAD 11|ACAD-11|Acad11|ACD11_HUMAN|Acyl CoA dehydrogenase family member 11|Acyl Coenzyme A dehydrogenase family member 11|Acyl-CoA dehydrogenase family member 11|AI987948|FLJ12592|MGC150619|Putative acyl CoA dehydrogenase|RGD1306270
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000
Type:
Antigen: ACAD11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human