Order Entry
United States
ContactUsLinkComponent
Anti-KCNMB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-KCNMB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-KCNMB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77824-590
  • 77824-589
  • Antigen Symbol:
  • KCNMB1
  • KCNMB1
  • Antigen Name:
  • Calcium-activated potassium channel subunit beta-1 protein (40-130AA)
  • Calcium-activated potassium channel subunit beta-1 protein (40-130AA)
  • Antigen Synonyms:
  • BK channel beta subunit|BK channel subunit beta 1|BK channel subunit beta-1|BKbeta|BKbeta1|Calcium activated potassium channel subfamily M subunit beta 1|Calcium activated potassium channel subunit beta 1|Calcium activated potassium channel subunit beta|Calcium-activated potassium channel|Calcium-activated potassium channel subunit beta|Calcium-activated potassium channel subunit beta-1|Charybdotoxin receptor subunit beta 1|Charybdotoxin receptor subunit beta-1|Hbeta1|hslo beta|K(VCA)beta 1|K(VCA)beta|K(VCA)beta-1|KCMB1_HUMAN|KCNMB 1|Kcnmb1|Large conductance Ca2+ activated K+ channel beta 1 subunit|Maxi K channel beta subunit|Maxi K channel subunit beta 1|Maxi K channel subunit beta-1|Potassium large conductance calcium activated channel subfamily M beta member 1|Slo beta 1|SLO beta|Slo-beta|Slo-beta-1|subfamily M subunit beta-1
  • BK channel beta subunit|BK channel subunit beta 1|BK channel subunit beta-1|BKbeta|BKbeta1|Calcium activated potassium channel subfamily M subunit beta 1|Calcium activated potassium channel subunit beta 1|Calcium activated potassium channel subunit beta|Calcium-activated potassium channel|Calcium-activated potassium channel subunit beta|Calcium-activated potassium channel subunit beta-1|Charybdotoxin receptor subunit beta 1|Charybdotoxin receptor subunit beta-1|Hbeta1|hslo beta|K(VCA)beta 1|K(VCA)beta|K(VCA)beta-1|KCMB1_HUMAN|KCNMB 1|Kcnmb1|Large conductance Ca2+ activated K+ channel beta 1 subunit|Maxi K channel beta subunit|Maxi K channel subunit beta 1|Maxi K channel subunit beta-1|Potassium large conductance calcium activated channel subfamily M beta member 1|Slo beta 1|SLO beta|Slo-beta|Slo-beta-1|subfamily M subunit beta-1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q16558
  • Q16558
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together