Anti-ST3GAL3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ST3GAL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-642
- 77821-641
- Antigen Symbol:
- ST3GAL3
- ST3GAL3
- Antigen Name:
- CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase protein (29-375AA)
- CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase protein (29-375AA)
- Antigen Synonyms:
- 3 sialyltransferase|3(4) GlcNAc alpha-2|3-sialyltransferase 3|3-sialyltransferase|3-ST 3|4-galactoside alpha-2|4GlcNAc alpha 2 3 sialyltransferase|Alpha 2 3 sialyltransferase II|Alpha 2 3 sialyltransferase III|Alpha 2 3 ST 3|Alpha 2|Beta galactoside alpha 3 sialyltransferase 3|Beta-galactoside alpha-2|CMP N acetylneuraminate beta 1 4 galactoside alpha 2 3 sialyltransferase|CMP-N-acetylneuraminate-beta-1|EC 2.4.99.6|Gal beta 1 3|Gal beta 1 3(4) GlcNAc alpha 2 3 sialyltransferase|Gal beta 1 3(4)GlcNAc alpha 2 3 sialyltransferase|Gal beta-1|N acetyllactosaminide alpha 2 3 sialyltransferase|N-acetyllactosaminide alpha-2|OTTHUMP00000008820|OTTHUMP00000008821|OTTHUMP00000008822|OTTHUMP00000008823|Sialyltransferase 6 (N acetyllacosaminide alpha 2 3 sialyltransferase)|Sialyltransferase 6|SIAT6|SIAT6_HUMAN|ST3 beta galactoside alpha 2 3 sialyltransferase 3|ST3 beta galactoside alpha 2,3 sialyltransferase 3|ST3Gal III|St3gal3|ST3GALII|ST3GalIII|ST3N
- 3 sialyltransferase|3(4) GlcNAc alpha-2|3-sialyltransferase 3|3-sialyltransferase|3-ST 3|4-galactoside alpha-2|4GlcNAc alpha 2 3 sialyltransferase|Alpha 2 3 sialyltransferase II|Alpha 2 3 sialyltransferase III|Alpha 2 3 ST 3|Alpha 2|Beta galactoside alpha 3 sialyltransferase 3|Beta-galactoside alpha-2|CMP N acetylneuraminate beta 1 4 galactoside alpha 2 3 sialyltransferase|CMP-N-acetylneuraminate-beta-1|EC 2.4.99.6|Gal beta 1 3|Gal beta 1 3(4) GlcNAc alpha 2 3 sialyltransferase|Gal beta 1 3(4)GlcNAc alpha 2 3 sialyltransferase|Gal beta-1|N acetyllactosaminide alpha 2 3 sialyltransferase|N-acetyllactosaminide alpha-2|OTTHUMP00000008820|OTTHUMP00000008821|OTTHUMP00000008822|OTTHUMP00000008823|Sialyltransferase 6 (N acetyllacosaminide alpha 2 3 sialyltransferase)|Sialyltransferase 6|SIAT6|SIAT6_HUMAN|ST3 beta galactoside alpha 2 3 sialyltransferase 3|ST3 beta galactoside alpha 2,3 sialyltransferase 3|ST3Gal III|St3gal3|ST3GALII|ST3GalIII|ST3N
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q11203
- Q11203
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together