Anti-TIMELESS Rabbit Polyclonal Antibody
Catalog # 77802-557
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Tof1 homolog
- Antigen Symbol:TIMELESS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UNS1
- Antigen Synonyms:FLJ12640|FLJ20714|hTIM|Protein timeless homolog|TIM|TIM_HUMAN|TIM1|Timeless|timeless circadian clock 1|timeless circadian clock|timeless homolog|TIMELESS1|Tof1 homolog
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TIMELESS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human