Anti-TCEAL9 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TCEAL9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-762
- 77842-761
- Antigen Symbol:
- TCEAL9
- TCEAL9
- Antigen Name:
- WW domain-binding protein 5 protein (1-104AA)
- WW domain-binding protein 5 protein (1-104AA)
- Antigen Synonyms:
- TCEAL9|WBP5|Transcription elongation factor A protein-like 9|TCEA-like protein 9|Transcription elongation factor S-II protein-like 9|WW domain-binding protein 5|WBP-5
- TCEAL9|WBP5|Transcription elongation factor A protein-like 9|TCEA-like protein 9|Transcription elongation factor S-II protein-like 9|WW domain-binding protein 5|WBP-5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHQ7
- Q9UHQ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together