Anti-SEPT3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SEPT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-056
- 77799-055
- Antigen Symbol:
- SEPT3
- SEPT3
- Antigen Name:
- Neuronal-specific septin-3 protein (238-358AA)
- Neuronal-specific septin-3 protein (238-358AA)
- Antigen Synonyms:
- bK250D10.3|MGC133218|Neuronal specific septin 3|Neuronal-specific septin-3|OTTHUMP00000028742|SEP 3|SEP3|SEPT 3|sept3|SEPT3 protein|SEPT3_HUMAN|Septin3
- bK250D10.3|MGC133218|Neuronal specific septin 3|Neuronal-specific septin-3|OTTHUMP00000028742|SEP 3|SEP3|SEPT 3|sept3|SEPT3 protein|SEPT3_HUMAN|Septin3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UH03
- Q9UH03
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together