Anti-IL21R Rabbit Recombinant Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: IL21R
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-709
- 77835-708
- Antigen Symbol:
- IL21R
- IL21R
- Antigen Name:
- Interleukin-21 receptor protein (20-240AA)
- Interleukin-21 receptor protein (20-240AA)
- Antigen Synonyms:
- CD360|IL 21R|IL-21 receptor|IL-21R|IL21 receptor|IL21R|IL21R_HUMAN|Interleukin 21 receptor|Interleukin-21 receptor|MGC10967|NILR|Novel interleukin receptor
- CD360|IL 21R|IL-21 receptor|IL-21R|IL21 receptor|IL21R|IL21R_HUMAN|Interleukin 21 receptor|Interleukin-21 receptor|MGC10967|NILR|Novel interleukin receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBE5
- Q9HBE5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together