Order Entry
United States
ContactUsLinkComponent
Anti-ALYREF Rabbit Recombinant Polyclonal Antibody
Anti-ALYREF Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ALYREF Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77832-701
  • 77832-700
  • Antigen Symbol:
  • ALYREF
  • ALYREF
  • Antigen Name:
  • THO complex subunit 4 protein (186-240AA)
  • THO complex subunit 4 protein (186-240AA)
  • Antigen Synonyms:
  • Ally of AML-1 and LEF-1|Ally of AML1 and LEF1|ALY|ALY/REF|Aly/REF export factor|BEF|bZIP enhancing factor|bZIP-enhancing factor BEF|REF|THO complex 4|THO complex subunit 4|Tho4|thoc4|THOC4_HUMAN|Transcriptional coactivator Aly/REF|Transcriptional coactivator
  • Ally of AML-1 and LEF-1|Ally of AML1 and LEF1|ALY|ALY/REF|Aly/REF export factor|BEF|bZIP enhancing factor|bZIP-enhancing factor BEF|REF|THO complex 4|THO complex subunit 4|Tho4|thoc4|THOC4_HUMAN|Transcriptional coactivator Aly/REF|Transcriptional coactivator
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q86V81
  • Q86V81
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together