Anti-RASSF4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RASSF4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-585
- 77840-586
- Antigen Symbol:
- RASSF4
- RASSF4
- Antigen Name:
- Ras association domain-containing protein 4 protein (65-180AA)
- Ras association domain-containing protein 4 protein (65-180AA)
- Antigen Synonyms:
- 3830411C14Rik|AD037|AI586137|MGC105606|MGC44914|OTTHUMP00000019493|Ras association (RalGDS/AF 6) domain family 4|Ras association (RalGDS/AF 6) domain family member 4|Ras association domain containing protein 4|Ras association domain family 4|Ras association domain-containing protein 4|RASF4_HUMAN|RASSF 4|RP11-285G1.4|Tumor suppressor RASSF4
- 3830411C14Rik|AD037|AI586137|MGC105606|MGC44914|OTTHUMP00000019493|Ras association (RalGDS/AF 6) domain family 4|Ras association (RalGDS/AF 6) domain family member 4|Ras association domain containing protein 4|Ras association domain family 4|Ras association domain-containing protein 4|RASF4_HUMAN|RASSF 4|RP11-285G1.4|Tumor suppressor RASSF4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2L5
- Q9H2L5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together