Anti-TNXB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TNXB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-297
- 77803-297
- Antigen Symbol:
- TNXB
- TNXB
- Antigen Name:
- Tenascin-X protein (3933-4242AA)
- Tenascin-X protein (3933-4242AA)
- Antigen Synonyms:
- EDS3|Ehlers Danlos like syndrome|Growth inhibiting protein 45|Hexabrachion like protein|Hexabrachion-like protein|HXBL|NXB2|Tenascin X|Tenascin X precursor|Tenascin XB|Tenascin XB1|Tenascin XB2|Tenascin-X|TENX|TENX_HUMAN|TN X|TN-X|TNX|TNXB|TNXB1|TNXB2|TNXBS|VUR8|XB|XBS
- EDS3|Ehlers Danlos like syndrome|Growth inhibiting protein 45|Hexabrachion like protein|Hexabrachion-like protein|HXBL|NXB2|Tenascin X|Tenascin X precursor|Tenascin XB|Tenascin XB1|Tenascin XB2|Tenascin-X|TENX|TENX_HUMAN|TN X|TN-X|TNX|TNXB|TNXB1|TNXB2|TNXBS|VUR8|XB|XBS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P22105
- P22105
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together