Anti-UQCRFS1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: UQCRFS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77807-845
- 77807-844
- Antigen Symbol:
- UQCRFS1
- UQCRFS1
- Antigen Name:
- Cytochrome b-c1 complex subunit Rieske, mitochondrial protein (79-274AA)
- Cytochrome b-c1 complex subunit Rieske, mitochondrial protein (79-274AA)
- Antigen Synonyms:
- Complex III subunit 5| Complex III subunit IX| Cytochrome b c1 complex subunit Rieske mitochondrial| Cytochrome b-c1 complex subunit 11| Cytochrome b-c1 complex subunit 5| Cytochrome b6f complex iron sulfur subunit, chloroplastic| petC| PGR1| Plastohydroquinone:plastocyanin oxidoreductase iron sulfur protein| Proton gradient regulation protein 1| Rieske iron sulfur protein| Rieske iron-sulfur protein| RIP1| RIS1| RISP| Ubiquinol cytochrome C reductase rieske iron sulphur| Ubiquinol cytochrome c reductase Rieske iron-sulfur polypeptide 1| Ubiquinol-cytochrome c reductase 8 kDa protein| Ubiquinol-cytochrome c reductase iron-sulfur subunit| UCRI_HUMAN| UQCR5| UQCRFS1
- Complex III subunit 5| Complex III subunit IX| Cytochrome b c1 complex subunit Rieske mitochondrial| Cytochrome b-c1 complex subunit 11| Cytochrome b-c1 complex subunit 5| Cytochrome b6f complex iron sulfur subunit, chloroplastic| petC| PGR1| Plastohydroquinone:plastocyanin oxidoreductase iron sulfur protein| Proton gradient regulation protein 1| Rieske iron sulfur protein| Rieske iron-sulfur protein| RIP1| RIS1| RISP| Ubiquinol cytochrome C reductase rieske iron sulphur| Ubiquinol cytochrome c reductase Rieske iron-sulfur polypeptide 1| Ubiquinol-cytochrome c reductase 8 kDa protein| Ubiquinol-cytochrome c reductase iron-sulfur subunit| UCRI_HUMAN| UQCR5| UQCRFS1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P47985
- P47985
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together