Anti-SFTPC Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SFTPC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-385
- 77799-384
- Antigen Symbol:
- SFTPC
- SFTPC
- Antigen Name:
- Pulmonary surfactant-associated protein C protein (24-58AA)
- Pulmonary surfactant-associated protein C protein (24-58AA)
- Antigen Synonyms:
- BRICD6|BRICHOS domain containing 6|PSP C|PSPC|PSPC_HUMAN|Pulmonary surfactant apoprotein 2|Pulmonary surfactant apoprotein PSP C|pulmonary surfactant apoprotein-2 SP-C|Pulmonary surfactant associated protein C|Pulmonary surfactant associated proteolipid SPL pVal|Pulmonary surfactant associated proteolipid SPL(Val)|Pulmonary surfactant protein SP5|Pulmonary surfactant-associated protein C|Pulmonary surfactant-associated proteolipid SPL(Val)|SFTP 2|SFTP2|SFTPC surfactant pulmonary associated protein C|SMDP2|SP 5|SP C|SP-C|SP5|SPC|Surfactant associated protein pulmonary 2|Surfactant protein c|Surfactant proteolipid SPL-pVal|Surfactant pulmonary associated protein C
- BRICD6|BRICHOS domain containing 6|PSP C|PSPC|PSPC_HUMAN|Pulmonary surfactant apoprotein 2|Pulmonary surfactant apoprotein PSP C|pulmonary surfactant apoprotein-2 SP-C|Pulmonary surfactant associated protein C|Pulmonary surfactant associated proteolipid SPL pVal|Pulmonary surfactant associated proteolipid SPL(Val)|Pulmonary surfactant protein SP5|Pulmonary surfactant-associated protein C|Pulmonary surfactant-associated proteolipid SPL(Val)|SFTP 2|SFTP2|SFTPC surfactant pulmonary associated protein C|SMDP2|SP 5|SP C|SP-C|SP5|SPC|Surfactant associated protein pulmonary 2|Surfactant protein c|Surfactant proteolipid SPL-pVal|Surfactant pulmonary associated protein C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11686
- P11686
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together