Anti-STX2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: STX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77822-965
- 77822-966
- Antigen Symbol:
- STX2
- STX2
- Antigen Name:
- Synthetic peptide of STX2
- Synthetic peptide of STX2
- Antigen Synonyms:
- EPIM|Epimorphin|EPM|MGC51014|OTTHUMP00000238636|OTTHUMP00000238637|Stx2|STX2_HUMAN|STX2A|STX2B|STX2C|Syntaxin 2A|Syntaxin 2B|Syntaxin 2C|Syntaxin-2
- EPIM|Epimorphin|EPM|MGC51014|OTTHUMP00000238636|OTTHUMP00000238637|Stx2|STX2_HUMAN|STX2A|STX2B|STX2C|Syntaxin 2A|Syntaxin 2B|Syntaxin 2C|Syntaxin-2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32856
- P32856
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together