Anti-VCPIP1 Rabbit Polyclonal Antibody
77804-306
New Product
- Antigen Name:Deubiquitinating protein VCIP135
- Antigen Symbol:VCPIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96JH7
- Antigen Synonyms:Deubiquitinating protein VCIP135| DUBA3| KIAA1850| Valosin containing protein (p97)/p47 complex interacting protein 1| Valosin containing protein p97/p47 complex interacting protein 1| Valosin containing protein p97/p47 complex interacting protein p135| Valosin-containing protein p97/p47 complex-interacting protein 1| Valosin-containing protein p97/p47 complex-interacting protein p135| VCIP1_HUMAN| VCIP135| VCP/p47 complex-interacting 135-kDa protein| Vcpip1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: VCPIP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat