Anti-TADA2A Rabbit Polyclonal Antibody
Catalog # 77801-798
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:TADA2L
- Antigen Symbol:TADA2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O75478
- Antigen Synonyms:TADA2A|TADA2L|KL04P|Transcriptional adapter 2-alpha|Transcriptional adapter 2-like|ADA2-like protein
- Storage Buffer:PBS, 0.1% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TADA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat