Anti-CARMIL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CARMIL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-908
- 77827-909
- Antigen Symbol:
- CARMIL1
- CARMIL1
- Antigen Name:
- F-actin-uncapping protein LRRC16A protein (1184-1295AA)
- F-actin-uncapping protein LRRC16A protein (1184-1295AA)
- Antigen Synonyms:
- Acan 125|Capping protein|Arp2/3|and myosin I linker|CARMIL homolog|leucine rich repeat containing 16|Leucine-rich repeat-containing protein 16A|LR16A_HUMAN|Lrrc16 protein|LRRC16A|p116
- Acan 125|Capping protein|Arp2/3|and myosin I linker|CARMIL homolog|leucine rich repeat containing 16|Leucine-rich repeat-containing protein 16A|LR16A_HUMAN|Lrrc16 protein|LRRC16A|p116
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5VZK9
- Q5VZK9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together