Anti-PPP2R2D Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPP2R2D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-673
- 77828-672
- Antigen Symbol:
- PPP2R2D
- PPP2R2D
- Antigen Name:
- Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform protein (1-192AA)
- Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform protein (1-192AA)
- Antigen Synonyms:
- 2ABD_HUMAN|MDS026|PP2A subunit B isoform B55-delta|PP2A subunit B isoform delta|PP2A subunit B isoform PR55-delta|PP2A subunit B isoform R2-delta|PPP2R2D|Protein phosphatase 2 regulatory subunit B delta|Protein phosphatase 2 regulatory subunit B delta isoform|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform
- 2ABD_HUMAN|MDS026|PP2A subunit B isoform B55-delta|PP2A subunit B isoform delta|PP2A subunit B isoform PR55-delta|PP2A subunit B isoform R2-delta|PPP2R2D|Protein phosphatase 2 regulatory subunit B delta|Protein phosphatase 2 regulatory subunit B delta isoform|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q66LE6
- Q66LE6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together