Anti-ADGRG3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADGRG3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-039
- 77832-040
- Antigen Symbol:
- ADGRG3
- ADGRG3
- Antigen Name:
- Adhesion G protein-coupled receptor G3 protein (98-244AA)
- Adhesion G protein-coupled receptor G3 protein (98-244AA)
- Antigen Synonyms:
- EGF-TM7-like|G protein coupled receptor 97|G-protein coupled receptor PGR26|GPR97|GPR97_HUMAN|Pb 99|Pb99|PGR 26|PGR26|Probable G-protein coupled receptor 97
- EGF-TM7-like|G protein coupled receptor 97|G-protein coupled receptor PGR26|GPR97|GPR97_HUMAN|Pb 99|Pb99|PGR 26|PGR26|Probable G-protein coupled receptor 97
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86Y34
- Q86Y34
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together