Anti-COQ3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: COQ3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-166
- 77841-167
- Antigen Symbol:
- COQ3
- COQ3
- Antigen Name:
- Ubiquinone biosynthesis O-methyltransferase, mitochondrial protein (271-369AA)
- Ubiquinone biosynthesis O-methyltransferase, mitochondrial protein (271-369AA)
- Antigen Synonyms:
- COQ3|UG0215E05Ubiquinone biosynthesis O-methyltransferase|mitochondrial|3-demethylubiquinol 3-O-methyltransferase|EC 2.1.1.64|Polyprenyldihydroxybenzoate methyltransferase|EC 2.1.1.114
- COQ3|UG0215E05Ubiquinone biosynthesis O-methyltransferase|mitochondrial|3-demethylubiquinol 3-O-methyltransferase|EC 2.1.1.64|Polyprenyldihydroxybenzoate methyltransferase|EC 2.1.1.114
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZJ6
- Q9NZJ6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together