Anti-SCGB2A1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SCGB2A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-757
- 77798-756
- Antigen Symbol:
- SCGB2A1
- SCGB2A1
- Antigen Name:
- Mammaglobin-B protein (19-95AA)
- Mammaglobin-B protein (19-95AA)
- Antigen Synonyms:
- Lacryglobin|LIPHC|Lipophilin C|Lipophilin-C|LPHC|LPNC|Mammaglobin 2|Mammaglobin-2|Mammaglobin-B|MGB2|MGC71973|Scgb2a1|Secretoglobin family 2A member 1|Secretoglobin|family 2A|member 1|SG2A1_HUMAN|UGB3
- Lacryglobin|LIPHC|Lipophilin C|Lipophilin-C|LPHC|LPNC|Mammaglobin 2|Mammaglobin-2|Mammaglobin-B|MGB2|MGC71973|Scgb2a1|Secretoglobin family 2A member 1|Secretoglobin|family 2A|member 1|SG2A1_HUMAN|UGB3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75556
- O75556
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together