Anti-BNIP1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: BNIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-166
- 77825-165
- Antigen Symbol:
- BNIP1
- BNIP1
- Antigen Name:
- Vesicle transport protein SEC20 protein (1-200AA)
- Vesicle transport protein SEC20 protein (1-200AA)
- Antigen Synonyms:
- BCL2 adenovirus E1B 19kD interacting protein 1| BCL2/adenovirus E1B 19 kDa protein-interacting protein 1| BNIP1| MGC41600| Nip1| OTTHUMP00000161077| OTTHUMP00000161078| OTTHUMP00000161079| OTTHUMP00000223580| sec20| SEC20_HUMAN| SEC20L| Transformation-related gene 8 protein| TRG-8| TRG8| Vesicle transport protein SEC20
- BCL2 adenovirus E1B 19kD interacting protein 1| BCL2/adenovirus E1B 19 kDa protein-interacting protein 1| BNIP1| MGC41600| Nip1| OTTHUMP00000161077| OTTHUMP00000161078| OTTHUMP00000161079| OTTHUMP00000223580| sec20| SEC20_HUMAN| SEC20L| Transformation-related gene 8 protein| TRG-8| TRG8| Vesicle transport protein SEC20
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12981
- Q12981
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together