Anti-POU3F2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POU3F2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-892
- 77794-893
- Antigen Symbol:
- POU3F2
- POU3F2
- Antigen Name:
- POU domain, class 3, transcription factor 2 protein (1-443AA)
- POU domain, class 3, transcription factor 2 protein (1-443AA)
- Antigen Synonyms:
- Brain 2|Brain specific homeobox/POU domain protein 2|Brain-2|Brain-specific homeobox/POU domain protein 2|BRN 2|Brn-2|BRN2|N Oct 3|N OCT 3 Gene|N Oct3|Nervous system specific octamer binding transcription factor|Nervous system-specific octamer-binding transcription factor N-Oct-3|OCT 7|Oct-7|OCT7|Octamer Binding Transcription Factor 7|Octamer-binding protein 7|Octamer-binding transcription factor 7|OTF 7|OTF-7|OTF7|PO3F2_HUMAN|POU class 3 homeobox 2,|POU domain class 3 transcription factor 2|POU domain, class 3, transcription factor 2|Pou3f2|POUF 3|POUF3|Protein Brn 2
- Brain 2|Brain specific homeobox/POU domain protein 2|Brain-2|Brain-specific homeobox/POU domain protein 2|BRN 2|Brn-2|BRN2|N Oct 3|N OCT 3 Gene|N Oct3|Nervous system specific octamer binding transcription factor|Nervous system-specific octamer-binding transcription factor N-Oct-3|OCT 7|Oct-7|OCT7|Octamer Binding Transcription Factor 7|Octamer-binding protein 7|Octamer-binding transcription factor 7|OTF 7|OTF-7|OTF7|PO3F2_HUMAN|POU class 3 homeobox 2,|POU domain class 3 transcription factor 2|POU domain, class 3, transcription factor 2|Pou3f2|POUF 3|POUF3|Protein Brn 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20265
- P20265
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together