Anti-EIPR1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EIPR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-762
- 77826-761
- Antigen Symbol:
- EIPR1
- EIPR1
- Antigen Name:
- EARP and GARP complex-interacting protein 1 protein (201-387AA)
- EARP and GARP complex-interacting protein 1 protein (201-387AA)
- Antigen Synonyms:
- EIPR1|TSSC1EARP and GARP complex-interacting protein 1|Endosome-associated recycling protein-interacting protein|Golgi-associated retrograde protein-interacting protein|Tumor-suppressing STF cDNA 1 protein|Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein
- EIPR1|TSSC1EARP and GARP complex-interacting protein 1|Endosome-associated recycling protein-interacting protein|Golgi-associated retrograde protein-interacting protein|Tumor-suppressing STF cDNA 1 protein|Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53HC9
- Q53HC9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together