Anti-UBE2L3 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2L3
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-863
- 77811-863
- Antigen Symbol:
- UBE2L3
- UBE2L3
- Antigen Name:
- Ubiquitin-conjugating enzyme E2 L3 protein (1-154AA)
- Ubiquitin-conjugating enzyme E2 L3 protein (1-154AA)
- Antigen Synonyms:
- E2 F1|L UBC|L-UBC|UB2L3_HUMAN|UBCE7|UbcH7|UbcM4|Ube2l3|Ubiquitin carrier protein L3|Ubiquitin conjugating enzyme E2 L3|Ubiquitin protein ligase L3|Ubiquitin-conjugating enzyme E2 L3|Ubiquitin-conjugating enzyme E2-F1|Ubiquitin-protein ligase L3
- E2 F1|L UBC|L-UBC|UB2L3_HUMAN|UBCE7|UbcH7|UbcM4|Ube2l3|Ubiquitin carrier protein L3|Ubiquitin conjugating enzyme E2 L3|Ubiquitin protein ligase L3|Ubiquitin-conjugating enzyme E2 L3|Ubiquitin-conjugating enzyme E2-F1|Ubiquitin-protein ligase L3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P68036
- P68036
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together