Anti-PTOV1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PTOV1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-045
- 77832-046
- Antigen Symbol:
- PTOV1
- PTOV1
- Antigen Name:
- Prostate tumor-overexpressed gene 1 protein (2-137AA)
- Prostate tumor-overexpressed gene 1 protein (2-137AA)
- Antigen Synonyms:
- ACID2|PP642|UNQ6127/PRO20092|Prostate tumor-overexpressed gene 1 protein|PTOV-1|Activator interaction domain-containing protein 2
- ACID2|PP642|UNQ6127/PRO20092|Prostate tumor-overexpressed gene 1 protein|PTOV-1|Activator interaction domain-containing protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86YD1
- Q86YD1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together