Anti-DHX36 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DHX36
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-514
- 77835-513
- Antigen Symbol:
- DHX36
- DHX36
- Antigen Name:
- ATP-dependent RNA helicase DHX36 protein (68-181AA)
- ATP-dependent RNA helicase DHX36 protein (68-181AA)
- Antigen Synonyms:
- DDX36|DEAH box protein 36|Dhx36|DHX36_HUMAN|EC 3.6.1.-|KIAA1488|MLE like protein 1|MLE-like protein 1|MLEL1|Probable ATP dependent RNA helicase DHX36|Probable ATP-dependent RNA helicase DHX36|RHAU|RNA helicase associated with AU rich element ARE|RNA helicase associated with AU-rich element ARE
- DDX36|DEAH box protein 36|Dhx36|DHX36_HUMAN|EC 3.6.1.-|KIAA1488|MLE like protein 1|MLE-like protein 1|MLEL1|Probable ATP dependent RNA helicase DHX36|Probable ATP-dependent RNA helicase DHX36|RHAU|RNA helicase associated with AU rich element ARE|RNA helicase associated with AU-rich element ARE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2U1
- Q9H2U1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together