Anti-PTDSS1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77807-651
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Phosphatidylserine synthase 1 protein (1-35AA)
- Antigen Symbol:PTDSS1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P48651
- Antigen Synonyms:PTDSS1|KIAA0024|PSSA|Phosphatidylserine synthase 1|PSS-1|PtdSer synthase 1|EC 2.7.8.29|Serine-exchange enzyme I
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PTDSS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human