Anti-RAD23A Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAD23A
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-987
- 77806-988
- Antigen Symbol:
- RAD23A
- RAD23A
- Antigen Name:
- UV excision repair protein RAD23 homolog A protein (2-201AA)
- UV excision repair protein RAD23 homolog A protein (2-201AA)
- Antigen Synonyms:
- hHR 23A|hHR23A|HR23A|MGC111083|RAD 23a|RAD23 homolog A (S. cerevisiae)|RAD23 homolog A|RAD23 yeast homolog A|RAD23A|RD23A_HUMAN|UV excision repair protein RAD23|UV excision repair protein RAD23 homolog A
- hHR 23A|hHR23A|HR23A|MGC111083|RAD 23a|RAD23 homolog A (S. cerevisiae)|RAD23 homolog A|RAD23 yeast homolog A|RAD23A|RD23A_HUMAN|UV excision repair protein RAD23|UV excision repair protein RAD23 homolog A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54725
- P54725
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together