Anti-TCF20 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TCF20
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-715
- 77842-716
- Antigen Symbol:
- TCF20
- TCF20
- Antigen Name:
- Transcription factor 20 protein (539-704AA)
- Transcription factor 20 protein (539-704AA)
- Antigen Synonyms:
- TCF20|KIAA0292|SPBP|Transcription factor 20|TCF-20|Nuclear factor SPBP|Protein AR1|Stromelysin-1 PDGF-responsive element-binding protein|SPRE-binding protein
- TCF20|KIAA0292|SPBP|Transcription factor 20|TCF-20|Nuclear factor SPBP|Protein AR1|Stromelysin-1 PDGF-responsive element-binding protein|SPRE-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UGU0
- Q9UGU0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together